4-(2-Hydroxyethoxy)benzenecarboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-(2-Hydroxyethoxy)benzoic acid, 4-(2-Hydroxy-ethoxy)-benzoic acid

OR962445-1G OR962445-5G
APOSOR962445-1GEA 114 EUR
APOSOR962445-1G APOSOR962445-5G
4-(2-Hydroxyethoxy)benzenecarboxylic acid 97%
4-(2-Hydroxyethoxy)benzenecarboxylic acid
Formel: C₉H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00020367
CAS-Nummer: 1711-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR