3-(3-Methoxyphenoxy)propionsäure 95%
Lieferant: Apollo Scientific
Synonyme:
3-(3-Methoxyphenoxy)propionic Acid
OR962463-1G
OR962463-5G
APOSOR962463-1GEA
62.5
EUR
APOSOR962463-1G
APOSOR962463-5G
3-(3-Methoxyphenoxy)propionsäure 95%
3-(3-Methoxyphenoxy)propionsäure
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,1999 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045943 CAS-Nummer: 49855-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...