4-Hydroxypyrimidine-5-carbonitrile 97%

Lieferant: Apollo Scientific

OR962470-1G OR962470-5G
APOSOR962470-1GEA 37.7 EUR
APOSOR962470-1G APOSOR962470-5G
4-Hydroxypyrimidine-5-carbonitrile 97%
4-Hydroxypyrimidine-5-carbonitrile
Formel: C₅H₃N₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 4774-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR