7-Azabicyclo[2.2.1]heptane hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
7-azabicyclo[2.2.1]heptane hydrochloride
OR962480-1G
OR962480-250MG
APOSOR962480-1GEA
137
EUR
APOSOR962480-1G
APOSOR962480-250MG
7-Azabicyclo[2.2.1]heptane hydrochloride 97%
7-Azabicyclo[2.2.1]heptane hydrochloride
Formel:
C₆H₁₂ClN Molecular Weight: 133.6200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154183 CAS-Nummer: 27514-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...