N-Methyl-3-nitropyridin-4-amine 98%
Lieferant: Apollo Scientific
Synonyme:
4-(Methylamino)-3-nitropyridine
OR962544-1G
OR962544-5G
APOSOR962544-1GEA
75.4
EUR
APOSOR962544-1G
APOSOR962544-5G
N-Methyl-3-nitropyridin-4-amine 98%
N-Methyl-3-nitropyridin-4-amine
Formel:
C₆H₇N₃O₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1633-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...