5-Brom-2-thiophencarbonitril 98%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-thiophenecarbonitrile, 5-Bromothiophene-2-carbonitrile, 2-Cyano-5-bromothiophene, 2-Bromo-5-cyanothiophene

OR962625-25G
APOSOR962625-25GEA 174 EUR
APOSOR962625-25G
5-Brom-2-thiophencarbonitril 98%
5-Brom-2-thiophencarbonitril
Formel: C₅H₂BrNS
Molecular Weight: 188,0500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974080
CAS-Nummer: 2160-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR