(4-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%
Lieferant: Apollo Scientific
OR962638-1G
OR962638-5G
APOSOR962638-1GEA
168
EUR
APOSOR962638-1G
APOSOR962638-5G
(4-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%
(4-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol
Formel:
C₁₁H₁₂N₂O Molecular Weight: 188,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864819 CAS-Nummer: 167758-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...