(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-1,2,3,4-Tetrahydronaphthoic acid
OR962670-10G
OR962670-1G
OR962670-5G
APOSOR962670-10GEA
387
EUR
APOSOR962670-10G
APOSOR962670-1G
APOSOR962670-5G
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoesäure 98%
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoesäure
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,22 g/mol Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368364 CAS-Nummer: 23357-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...