4-(1-Pyrrolidinyl)benzonitril 98%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Pyrrolidinyl)benzonitrile, 1-(4-Cyanophenyl)pyrrolidine

OR962672-25G OR962672-5G
APOSOR962672-25GEA 236 EUR
APOSOR962672-25G APOSOR962672-5G
4-(1-Pyrrolidinyl)benzonitril 98%
4-(Pyrrolidin-1-yl)benzonitrile
Formel: C₁₁H₁₂N₂
Molecular Weight: 172,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368512
CAS-Nummer: 10282-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR