3-(1H-Imidazol-1-yl)benzonitrile 95%

Lieferant: Apollo Scientific

OR962673-1G OR962673-25G OR962673-5G
APOSOR962673-1GEA 52.2 EUR
APOSOR962673-1G APOSOR962673-25G APOSOR962673-5G
3-(1H-Imidazol-1-yl)benzonitrile 95%
3-(1H-Imidazol-1-yl)benzonitrile
Formel: C₁₀H₇N₃
Molecular Weight: 169.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368519
CAS-Nummer: 25699-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR