3-(1H-Pyrazol-1-yl)benzonitril 95%

Lieferant: Apollo Scientific

OR962682-1G OR962682-25G OR962682-5G
APOSOR962682-1GEA 132 EUR
APOSOR962682-1G APOSOR962682-25G APOSOR962682-5G
3-(1H-Pyrazol-1-yl)benzonitril 95%
3-(1H-Pyrazol-1-yl)benzonitril
Formel: C₁₀H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07772807
CAS-Nummer: 25699-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR