2-(2-Methoxyethoxy)anilin 95%

Lieferant: Apollo Scientific

OR962692-10G OR962692-1G OR962692-5G
APOSOR962692-10GEA 1120 EUR
APOSOR962692-10G APOSOR962692-1G APOSOR962692-5G
2-(2-Methoxyethoxy)anilin 95%
2-(2-Methoxyethoxy)anilin
Formel: C₉H₁₃NO₂
Molecular Weight: 167,2050 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08144123
CAS-Nummer: 72806-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR