Propoxyacetic acid 98%
Lieferant: Apollo Scientific
OR962755-1G
OR962755-5G
APOSOR962755-1GEA
0
EUR
APOSOR962755-1G
APOSOR962755-5G
Propoxyacetic acid 98%
Propoxyacetic acid
Formel:
C₅H₁₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09744041 CAS-Nummer: 54497-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...