N-Methoxy-N-methylpropionamide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Methoxy-N-methylpropanamide

OR962763-1G OR962763-25G OR962763-5G
APOSOR962763-1GEA 125 EUR
APOSOR962763-1G APOSOR962763-25G APOSOR962763-5G
N-Methoxy-N-methylpropionamide 95%
N-Methoxy-N-methylpropionamide
Formel: C₅H₁₁NO₂
Dichte: 0,98 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD09833541
CAS-Nummer: 104863-65-2
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR