2-Brom-6-nitrobenzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-6-nitrophenyl)methanol

OR962824-1G OR962824-5G
APOSOR962824-1GEA 23.2 EUR
APOSOR962824-1G APOSOR962824-5G
2-Brom-6-nitrobenzylalkohol 97%
2-Brom-6-nitrobenzylalkohol
Formel: C₇H₆BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11036298
CAS-Nummer: 861106-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR