3-(Piperidin-4-yloxy)pyridine 98%
Lieferant: Apollo Scientific
OR962957-1G
OR962957-250MG
APOSOR962957-1GEA
435
EUR
APOSOR962957-1G
APOSOR962957-250MG
3-(Piperidin-4-yloxy)pyridine 98%
3-(Piperidin-4-yloxy)pyridine
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059774 CAS-Nummer: 310881-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...