N-Me-Ala-OtBu·HCl 98%
Lieferant: Apollo Scientific
Synonyme:
(S)-tert-Butyl 2-(methylamino)propanoate hydrochloride, N-Me-Ala-OtBu·HCl
OR962971-1G
OR962971-5G
APOSOR962971-1GEA
59.6
EUR
APOSOR962971-1G
APOSOR962971-5G
N-Me-Ala-OtBu·HCl 98%
N-Me-Ala-OtBu·HCl
Formel:
C₈H₁₈ClNO₂ Molecular Weight: 195,687 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08457605 CAS-Nummer: 103614-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...