6-Bromo-1-methyl-1H-indole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR9630-1G
OR9630-250MG
OR9630-5G
APOSOR9630-1GEA
87
EUR
APOSOR9630-1G
APOSOR9630-250MG
APOSOR9630-5G
6-Bromo-1-methyl-1H-indole 95+%
6-Bromo-1-methyl-1H-indole
Formel:
C₉H₈BrN Lagertemperatur: Kühlschrank |
MDL:
MFCD08543444 CAS-Nummer: 125872-95-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...