Ac-D-phenylglycinol 95%

Lieferant: Apollo Scientific

Synonyme: D(-)-Ac-α-phenylglycinol

OR963082-1G OR963082-5G
APOSOR963082-1GEA 226 EUR
APOSOR963082-1G APOSOR963082-5G
Ac-D-phenylglycinol 95%
Ac-D-phenylglycinol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038782
CAS-Nummer: 78761-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR