2-Brombenzo[b]thiophen
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-benzo[b]thiophene
OR9631-1G
OR9631-25G
OR9631-5G
APOSOR9631-1GEA
72.7
EUR
APOSOR9631-1G
APOSOR9631-25G
APOSOR9631-5G
2-Brombenzo[b]thiophen
2-Brombenzo[b]thiophen
Formel:
C₈H₅BrS Molecular Weight: 213.0943 g/mol Schmelzpunkt: 37…43 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08435846 CAS-Nummer: 5394-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...