Ac-Nle-OH 95%

Lieferant: Apollo Scientific

Synonyme: N-Ac-Nle-OH, (S)-2-Acetamidohexanoic acid, (2S)-2-Acetamidohexanoic acid

OR963188-25G OR963188-5G
APOSOR963188-25GEA 214 EUR
APOSOR963188-25G APOSOR963188-5G
Ac-Nle-OH 95%
Ac-Nle-OH
Formel: C₈H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00151961
CAS-Nummer: 15891-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR