Ac-DL-Pra-Oet 95%
Lieferant: Apollo Scientific
Synonyme: Ethyl 2-acetamidopent-4-ynoate
OR963272-1G OR963272-5G
APOSOR963272-1GEA 183 EUR
APOSOR963272-1G APOSOR963272-5G
Ac-DL-Pra-Oet 95%
Ac-DL-Pra-Oet
Formel: C₉H₁₃NO₃
Molecular Weight: 183,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237477
CAS-Nummer: 23235-05-4