4-(3-Thienyl)anilin 95%

Lieferant: Apollo Scientific

OR963398-1G OR963398-250MG
APOSOR963398-1GEA 270 EUR
APOSOR963398-1G APOSOR963398-250MG
4-(3-Thienyl)anilin 95%
4-(3-Thienyl)anilin
Formel: C₁₀H₉NS
Lagertemperatur: Raumtemperatur
MDL: MFCD06740168
CAS-Nummer: 834884-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR