4-Iodo-1H-pyrazol-5-amine 96%
Lieferant: Apollo Scientific
Synonyme:
3-Amino-4-iodo-1H-pyrazole, 4-Iodo-1H-pyrazol-3-amine, 4-Iodo-3-aminopyrazole
OR963404-1G
OR963404-5G
APOSOR963404-1GEA
201
EUR
APOSOR963404-1G
APOSOR963404-5G
4-Iodo-1H-pyrazol-5-amine 96%
4-Iodo-1H-pyrazol-5-amine
Formel:
C₃H₄IN₃ Molecular Weight: 208,99 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22422021 CAS-Nummer: 81542-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...