z-Tyr-OMe
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Z-Tyr-OMe, N-Benzyloxycarbonyl-L-tyrosine methyl ester
OR963410-100G
OR963410-25G
APOSOR963410-100GEA
94.4
EUR
APOSOR963410-100G
APOSOR963410-25G
z-Tyr-OMe
z-Tyr-OMe
Formel:
C₁₈H₁₉NO₅ Molecular Weight: 329,352 g/mol Schmelzpunkt: 93…97 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00038784 CAS-Nummer: 13512-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...