2-Allylcyclohexanon 95%

Lieferant: Apollo Scientific

OR963660-25G OR963660-5G
APOSOR963660-25GEA 828 EUR
APOSOR963660-25G APOSOR963660-5G
2-Allylcyclohexanon 95%
2-Allylcyclohexanon
Formel: C₉H₁₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00043548
CAS-Nummer: 94-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR