Boc-Cys(pMeOBzl)-OH 97+%

Lieferant: Apollo Scientific

Synonyme: Boc-Cys(Mob)-OH, N-Boc-S-(4-methoxybenzyl)-L-cysteine

OR963796-25G OR963796-100G
APOSOR963796-25GEA 74.1 EUR
APOSOR963796-25G APOSOR963796-100G
Boc-Cys(pMeOBzl)-OH 97+%
Boc-Cys(pMeOBzl)-OH
Formel: C₁₆H₂₃NO₅S
Molecular Weight: 341,422 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00038254
CAS-Nummer: 18942-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR