(2E)-3-(3,4-Dimethoxyphenyl)acrylonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dimethoxycinnamonitrile

OR963888-1G OR963888-5G
APOSOR963888-1GEA 161 EUR
APOSOR963888-1G APOSOR963888-5G
(2E)-3-(3,4-Dimethoxyphenyl)acrylonitrile 95%
(2E)-3-(3,4-Dimethoxyphenyl)acrylonitrile
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001931
CAS-Nummer: 6443-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR