2-Chlorothionicotinamide 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloropyridine-3-thiocarboxamide

OR963896-1G OR963896-5G
APOSOR963896-1GEA 132 EUR
APOSOR963896-1G APOSOR963896-5G
2-Chlorothionicotinamide 97%
2-Chlorothionicotinamide
Formel: C₆H₅ClN₂S
Molecular Weight: 172.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16659867
CAS-Nummer: 1240596-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR