2-Hydroxyacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: Phenacylhydroxyid, α-Hydroxyacetophenon, ω-Hydroxyacetophenon

OR963976-100G
APOSOR963976-100GEA 125 EUR
APOSOR963976-100G
2-Hydroxyacetophenon 95%
2-Hydroxyacetophenon
Formel: C₈H₈O₂
Molecular Weight: 136,15 g/mol
Siedepunkt: 126 °C/12mmHg
Schmelzpunkt: 83…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041829
CAS-Nummer: 582-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR