2-Methyl-3-pyrrolidin-1-ylpropan-1-ol 95%

Lieferant: Apollo Scientific

OR964016-1G OR964016-5G
APOSOR964016-1GEA 219 EUR
APOSOR964016-1G APOSOR964016-5G
2-Methyl-3-pyrrolidin-1-ylpropan-1-ol 95%
2-Methyl-3-pyrrolidin-1-ylpropan-1-ol
Formel: C₈H₁₇NO
Molecular Weight: 143,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061224
CAS-Nummer: 873376-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR