2-Methylchinoxalin 98%

Lieferant: Apollo Scientific

OR964038-100G OR964038-25G
APOSOR964038-100GEA 192 EUR
APOSOR964038-100G APOSOR964038-25G
2-Methylchinoxalin 98%
2-Methylquinoxaline
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006727
CAS-Nummer: 7251-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR