Z-D-MEAla-OH 97%
Lieferant: Apollo Scientific
Synonyme:
Z-N-Me-D-Ala-OH, Z-D-MeAla-OH, N-Benzyloxycarbonyl-N-methyl-D-alanine
OR964070-25G
APOSOR964070-25GEA
85.7
EUR
APOSOR964070-25G
Z-D-MEAla-OH 97%
Z-D-MEAla-OH
Formel:
C₁₂H₁₅NO₄ Molecular Weight: 237,255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153387 CAS-Nummer: 68223-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...