N-Benzoyl-D-alanine 98%

Lieferant: Apollo Scientific

Synonyme: Bz-D-Ala-OH

OR964163-25G OR964163-5G
APOSOR964163-25GEA 63.8 EUR
APOSOR964163-25G APOSOR964163-5G
N-Benzoyl-D-alanine 98%
N-Benzoyl-D-alanine
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063127
CAS-Nummer: 17966-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR