3-(1H-Imidazol-2-yl)pyridine 95%
Lieferant: Apollo Scientific
OR964166-1G
APOSOR964166-1GEA
58
EUR
APOSOR964166-1G
3-(1H-Imidazol-2-yl)pyridine 95%
3-(1H-Imidazol-2-yl)pyridine
Formel:
C₈H₇N₃ Molecular Weight: 145.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458369 CAS-Nummer: 13570-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...