D-Proline ethyl ester hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: H-D-Pro-OEt hydrochloride, (R)-Ethyl pyrrolidine-2-carboxylate hydrochloride

OR964185-25G OR964185-5G
APOSOR964185-25GEA 441 EUR
APOSOR964185-25G APOSOR964185-5G
D-Proline ethyl ester hydrochloride 97%
D-Proline ethyl ester hydrochloride
Formel: C₇H₁₄ClNO₂
Molecular Weight: 179.644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191003
CAS-Nummer: 131477-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR