D-Leucine ethyl ester hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
H-D-Leu-OEt.HCl, H-D-Leu-OEt·HCl
OR964283-25G
APOSOR964283-25GEA
58
EUR
APOSOR964283-25G
D-Leucine ethyl ester hydrochloride 95%
D-Leucine ethyl ester hydrochloride
Formel:
C₈H₁₇NO₂·HCl Molecular Weight: 195,687 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190936 CAS-Nummer: 73913-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...