Fmoc-Bpa-OH 97%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-4-benzoyl-L-phenylalanine, Fmoc-p-Bz-Phe-OH

OR964394-1G OR964394-5G
APOSOR964394-1GEA 68.3 EUR
APOSOR964394-1G APOSOR964394-5G
Fmoc-Bpa-OH 97%
Fmoc-Bpa-OH
Formel: C₃₁H₂₅NO₅
Molecular Weight: 491.543 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151937
CAS-Nummer: 117666-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR