(S)-3-[1-(Dimethylamino)ethyl]phenol 95%

Lieferant: Apollo Scientific

Synonyme: (S)-3-(1-(Dimethylamino)ethyl)phenol, NAP-226-90

OR964492-5G
APOSOR964492-5GEA 85.7 EUR
APOSOR964492-5G
(S)-3-[1-(Dimethylamino)ethyl]phenol 95%
(S)-3-[1-(Dimethylamino)ethyl]phenol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656491
CAS-Nummer: 139306-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR