Acetyl-L-threonine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ac-Thr-OH
OR964495-1G
OR964495-250MG
OR964495-5G
APOSOR964495-1GEA
0
EUR
APOSOR964495-1G
APOSOR964495-250MG
APOSOR964495-5G
Acetyl-L-threonine 97%
Acetyl-L-threonine
Formel:
C₆H₁₁NO₄ Molecular Weight: 161,157 g/mol Schmelzpunkt: 126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037810 CAS-Nummer: 17093-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...