Z-D-Norleucine 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-2-(((Benzyloxy)carbonyl)amino)hexanoic acid, Z-D-Nle-OH, Cbz-D-Nle-OH
OR964562-1G
OR964562-25G
OR964562-5G
APOSOR964562-1GEA
60.9
EUR
APOSOR964562-1G
APOSOR964562-25G
APOSOR964562-5G
Z-D-Norleucine 97%
Z-D-Norleucine
Formel:
C₁₄H₁₉NO₄ Molecular Weight: 265,309 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063169 CAS-Nummer: 15027-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...