6-Chloro-L-Tryptophan 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid, H-Trp(6-Cl)-OH

OR964662-1G OR964662-250MG OR964662-500MG OR964662-100MG
APOSOR964662-1GEA 690 EUR
APOSOR964662-1G APOSOR964662-250MG APOSOR964662-500MG APOSOR964662-100MG
6-Chloro-L-Tryptophan 97%
6-Chloro-L-Tryptophan
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683925
CAS-Nummer: 33468-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR