3-Amino-4-phenylbutanoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-β-HoPhe-OH, DL-β-Homophenylalanine

OR964701-1G OR964701-5G
APOSOR964701-1GEA 183 EUR
APOSOR964701-1G APOSOR964701-5G
3-Amino-4-phenylbutanoic acid 97%
3-Amino-4-phenylbutanoic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02090626
CAS-Nummer: 15099-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR