1,2-Phenylendiessigsäure 95%

Lieferant: Apollo Scientific

Synonyme: o-Phenylenediacetic acid, 2,2'-(1,2-Phenylene)diacetic acid

OR964730-25G
APOSOR964730-25GEA 56.7 EUR
APOSOR964730-25G
1,2-Phenylendiessigsäure 95%
1,2-Phenylendiessigsäure
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004329
CAS-Nummer: 7500-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR