5-Amino-3-(2-chlorphenyl)isoxazol 95%

Lieferant: Apollo Scientific

Synonyme: 3-(2-Chlorophenyl)isoxazole-5-amine

OR964757-1G OR964757-250MG
APOSOR964757-1GEA 348 EUR
APOSOR964757-1G APOSOR964757-250MG
5-Amino-3-(2-chlorphenyl)isoxazol 95%
5-Amino-3-(2-chlorophenyl)isoxazole
Formel: C₉H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06199356
CAS-Nummer: 27025-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR