3-Methyl-3H-imidazo[4,5-b]pyridine-6-carboxylic acid 95%

Lieferant: Apollo Scientific

OR964762-1G OR964762-250MG
APOSOR964762-1GEA 405 EUR
APOSOR964762-1G APOSOR964762-250MG
3-Methyl-3H-imidazo[4,5-b]pyridine-6-carboxylic acid 95%
3-Methyl-3H-imidazo[4,5-b]pyridine-6-carboxylic acid
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11857697
CAS-Nummer: 1138444-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR