3-Amino-2-thiophencarbonitril 95%
Lieferant: Apollo Scientific
OR964788-100MG
OR964788-1G
OR964788-250MG
APOSOR964788-100MGEA
161
EUR
APOSOR964788-100MG
APOSOR964788-1G
APOSOR964788-250MG
3-Amino-2-thiophencarbonitril 95%
3-Amino-2-thiophencarbonitril
Formel:
C₅H₄N₂S Lagertemperatur: Raumtemperatur |
CAS-Nummer:
56489-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...