5-(2-Furyl)nicotinic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9648-250MG
OR9648-1G
OR9648-5G
APOSOR9648-250MGEA
84.4
EUR
APOSOR9648-250MG
APOSOR9648-1G
APOSOR9648-5G
5-(2-Furyl)nicotinic acid 97%
5-(2-Furyl)nicotinic acid
Formel:
C₁₀H₇NO₃ Schmelzpunkt: 260,5…263 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410257 CAS-Nummer: 857283-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...