2-(2-Dimethylaminoethoxy)benzaldehyde 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9650-1G
OR9650-250MG
APOSOR9650-1GEA
132
EUR
APOSOR9650-1G
APOSOR9650-250MG
2-(2-Dimethylaminoethoxy)benzaldehyde 97%
2-(2-Dimethylaminoethoxy)benzaldehyde
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08146610 CAS-Nummer: 15182-06-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...