3-Cyclopentene-1-ol 96+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cyclopent-3-enol
OR965026-100G
OR965026-25G
APOSOR965026-100GEA
290
EUR
APOSOR965026-100G
APOSOR965026-25G
3-Cyclopentene-1-ol 96+%
3-Cyclopentene-1-ol
Formel:
C₅H₈O Molecular Weight: 84,12 g/mol Flammpunkt: 41,9 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798186 CAS-Nummer: 14320-38-8 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...